Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318060.1 | NP_001304989.1 | 2 | 710 | ||
NM_001318062.1 | NP_001304991.1 | 2 | 710 | |||
NM_006716.3 | NP_006707.1 | 2 | 710 | |||
GenBank mRNA | AB028069.1 | - | 2 | 723 | ||
AB028070.1 | - | 2 | 723 | |||
AF160249.1 | - | 2 | 422 | |||
AF160876.1 | - | 2 | 405 | |||
AK292569.1 | - | 2 | 452 | |||
AK302010.1 | - | 2 | 339 | |||
BC022811.1 | - | 2 | 307 | |||
BC036045.1 | - | 2 | 516 | 5273059 | ||
BC047693.1 | - | 2 | 391 | 5753567 |