Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019775.3 | NP_062749.2 | 4 | 436 | ||
GenBank mRNA | AA048136.1 | - | 4 | 436 | ||
AB021968.1 | - | 4 | 384 | |||
AF164524.1 | - | 4 | 451 | |||
AF186188.1 | - | 4 | 387 | |||
AK004045.1 | - | 4 | 409 | |||
BC089577.1 | - | 4 | 408 | 6766672 |