Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019775.3 | NP_062749.2 | 11 | 1183 | ||
GenBank mRNA | AB021968.1 | - | 11 | 1131 | ||
AF164524.1 | - | 11 | 1198 | |||
AF186188.1 | - | 11 | 1134 | |||
AK004045.1 | - | 11 | 1156 | |||
BC089577.1 | - | 11 | 1155 | 6766672 | ||
CO807338.1 | - | 3 | 217 |