Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019776.2 | NP_062750.2 | 2 | 375 | ||
GenBank mRNA | AB021491.1 | - | 2 | 332 | ||
AK034914.1 | - | 2 | 323 | |||
AK088028.1 | - | 2 | 334 | |||
AK133837.1 | - | 2 | 381 | |||
AK159837.1 | - | 2 | 340 | |||
AK161591.1 | - | 2 | 329 | |||
AK162432.1 | - | 2 | 316 | |||
AK167576.1 | - | 2 | 365 | |||
AK204079.1 | - | 1 | 41 | |||
BB613775.1 | - | 2 | 375 | |||
BC007126.1 | - | 2 | 322 | 3592548 | ||
BM944959.1 | - | 1 | 53 |