Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285805.1 | NP_001272734.1 | 9, 10 | 781 | ||
NM_019778.2 | NP_062752.2 | 7, 8 | 728 | |||
NM_181058.1 | NP_851401.1 | 5, 6 | 506 | |||
GenBank mRNA | AB221632.1 | - | 1, 2 | 193 | ||
AF185576.1 | - | 5, 6 | 551 | |||
AF194030.1 | - | 3, 4 | 343 | |||
AK005028.2 | - | 5, 6 | 507 | |||
AK052686.1 | - | 6 | 1055 | |||
AY028963.1 | - | 2, 3 | 310 | |||
BC031114.1 | - | 6, 7 | 614 | 4971586 | ||
BC056446.1 | - | 9, 10 | 753 | 5703319 |