Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285805.1 | NP_001272734.1 | 11 | 2901 | ||
NM_019778.2 | NP_062752.2 | 9 | 2848 | |||
NM_181058.1 | NP_851401.1 | 7 | 2626 | |||
GenBank mRNA | AF185576.1 | - | 7 | 2671 | ||
AF194030.1 | - | 5 | 2463 | |||
AK005028.2 | - | 7 | 2627 | |||
AY028963.1 | - | 4 | 2430 | |||
BC031114.1 | - | 8 | 2734 | 4971586 | ||
BC056446.1 | - | 11 | 2873 | 5703319 |