Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019785.2 | NP_062759.2 | 13 | 1231 | ||
GenBank mRNA | AF190797.1 | - | 13 | 1198 | ||
AK090043.1 | - | 13 | 1235 | |||
AK161642.1 | - | 13 | 1191 | |||
BC002227.1 | - | 13 | 1191 | 3491299 |