Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmeff2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019790.4 | NP_062764.1 | 10 | 1919 | ||
GenBank mRNA | AK029307.1 | - | 10 | 1919 | ||
AK135038.1 | - | 10 | 1906 | |||
BC034850.1 | - | 10 | 1895 | 1397175 |