Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019795.4 | NP_062769.2 | 3 | 760 | ||
GenBank mRNA | AB028861.1 | - | 3 | 208 | ||
AK011875.1 | - | 3 | 199 | |||
AK076032.1 | - | 3 | 231 | |||
AK145506.1 | - | 3 | 222 | |||
AK145739.1 | - | 3 | 222 | |||
AK166872.1 | - | 3 | 222 | |||
AY323828.1 | - | 3 | 196 | |||
BC023681.1 | - | 3 | 404 | 5343384 | ||
BC055729.1 | - | 3 | 759 | 6414499 |