Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019802.5 | NP_062776.1 | 7, 8 | 991 | ||
GenBank mRNA | AF087938.1 | - | 7, 8 | 966 | ||
AK033496.1 | - | 7, 8 | 918 | |||
AK036577.1 | - | 7, 8 | 965 | |||
AK135425.1 | - | 7, 8 | 978 | |||
AK165982.1 | - | 7, 8 | 966 | |||
AK206484.1 | - | 2, 3 | 193 | |||
BC138763.1 | - | 7, 8 | 916 | 8861785 | ||
BC138764.1 | - | 7, 8 | 916 | 8861786 |