Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019802.5 | NP_062776.1 | 15 | 2348 | ||
GenBank mRNA | AF087938.1 | - | 15 | 2323 | ||
AK030779.1 | - | 97 | ||||
AK033496.1 | - | 15 | 2275 | |||
AK036577.1 | - | 15 | 2322 | |||
AK135425.1 | - | 15 | 2335 | |||
AK165982.1 | - | 15 | 2323 | |||
BC138763.1 | - | 15 | 2273 | 8861785 | ||
BC138764.1 | - | 15 | 2273 | 8861786 |