Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285793.1 | NP_001272722.1 | 6 | 1210 | ||
NM_019804.4 | NP_062778.2 | 6 | 1143 | |||
GenBank mRNA | AF142672.1 | - | 6 | 1120 | ||
AF158746.1 | - | 6 | 1122 | |||
AK018658.1 | - | 6 | 1045 | |||
AK041692.1 | - | 6 | 1135 | |||
AK045602.1 | - | 6 | 1115 | |||
AK050005.1 | - | 6 | 1139 | |||
AK052013.1 | - | 6 | 1137 | |||
AK053603.1 | - | 6 | 1204 | |||
AK085368.1 | - | 6 | 1129 | |||
AK136042.1 | - | 2 | 315 | |||
BC013492.1 | - | 6 | 1106 | 4218187 | ||
BC031115.1 | - | 6 | 1195 | 5026168 |