Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019822.3 | NP_062796.2 | 2 | 265 | ||
GenBank mRNA | AF225959.1 | - | 2 | 181 | ||
AK010919.1 | - | 2 | 242 | |||
AK082084.1 | - | 2 | 248 | |||
AK143843.1 | - | 2 | 240 | |||
AK145474.1 | - | 2 | 227 | |||
AK145792.1 | - | 2 | 256 | |||
AK150091.1 | - | 2 | 218 | |||
AK166878.1 | - | 2 | 254 | |||
AK167085.1 | - | 1 | 44 | |||
AK207110.1 | - | 1 | 85 | |||
BC008974.1 | - | 2 | 265 | 3586698 | ||
BC019746.1 | - | 2 | 194 | 4188750 | ||
BC031517.1 | - | 2 | 208 | 4008987 |