Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019824.3 | NP_062798.1 | 2, 3 | 179 | ||
GenBank mRNA | AB038243.1 | - | 1, 2 | 115 | ||
AK003485.1 | - | 2, 3 | 210 | |||
AK005437.1 | - | 2, 3 | 198 | |||
AK171651.1 | - | 2, 3 | 194 | |||
BC013618.1 | - | 2, 3 | 179 | 4013537 | ||
BC054440.1 | - | 2, 3 | 146 | 6395280 | ||
BC055036.1 | - | 2, 3 | 146 |