Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019861.1 | NP_063914.1 | 5 | 656 | ||
GenBank mRNA | AF136280.1 | - | 5 | 557 | ||
AF197480.1 | - | 5 | 601 | |||
AJ131851.1 | - | 5 | 533 | |||
AK017474.1 | - | 5 | 542 | |||
AK075862.1 | - | 5 | 549 | |||
AK165416.1 | - | 4 | 716 | |||
BC004054.1 | - | 2 | 127 | |||
BC051131.1 | - | 4 | 775 | |||
BC058758.1 | - | 5 | 547 | 6492834 |