Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019861.1 | NP_063914.1 | 10 | 1227 | ||
GenBank mRNA | AF136280.1 | - | 10 | 1128 | ||
AF197480.1 | - | 10 | 1172 | |||
AJ131851.1 | - | 10 | 1104 | |||
AK017474.1 | - | 10 | 1110 | |||
AK075862.1 | - | 10 | 1120 | |||
BC004054.1 | - | 7 | 698 | |||
BC051131.1 | - | 9 | 1346 | |||
BC058758.1 | - | 10 | 1118 | 6492834 | ||
BC115438.1 | - | 4 | 347 |