Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019874.3 | NP_063927.1 | 3 | 980 | ||
GenBank mRNA | AF088983.1 | - | 4 | 1027 | ||
AF321322.1 | - | 4 | 1020 | |||
AK158535.1 | - | 4 | 1072 | |||
AK158972.1 | - | 4 | 1280 | |||
BC027782.1 | - | 3 | 3723 | |||
BC048902.1 | - | 4 | 1265 | 6396037 | ||
BC057087.1 | - | 3 | 980 | 5695243 |