Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suclg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019879.3 | NP_063932.2 | 4 | 554 | ||
GenBank mRNA | AF144101.2 | - | 4 | 387 | ||
AK002754.1 | - | 4 | 389 | |||
AK005080.1 | - | 4 | 403 | |||
AK088888.1 | - | 4 | 411 | |||
AK184184.1 | - | 4 | 330 | |||
BC011087.1 | - | 4 | 375 | 4012310 |