Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019911.2 | NP_064295.2 | 3 | 247 | ||
GenBank mRNA | AB476989.1 | - | 3 | 721 | ||
AB476990.1 | - | 3 | 607 | |||
AK132271.1 | - | 3 | 270 | |||
BC018390.1 | - | 3 | 247 | 4159877 | ||
U24493.1 | - | 3 | 234 |