Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2d2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019912.2 | NP_064296.1 | 7 | 1378 | ||
GenBank mRNA | AK005355.1 | - | 8 | 1327 | ||
AK136197.1 | - | 7 | 1317 | |||
AK139623.1 | - | 7 | 1344 | |||
AK147568.1 | - | 6 | 7064 | |||
AK148089.1 | - | 7 | 1200 | |||
AK162093.1 | - | 7 | 1184 | |||
AK172088.1 | - | 7 | 1205 | |||
AK218311.1 | - | 1 | 71 | |||
U62483.1 | - | 7 | 1316 |