Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019921.3 | NP_064305.2 | 4 | 563 | ||
GenBank mRNA | AK049399.1 | - | 4 | 532 | ||
AK051044.1 | - | 2 | 198 | |||
AK083255.1 | - | 4 | 532 | |||
AK162966.1 | - | 4 | 564 |