Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019921.3 | NP_064305.2 | 12 | 2002 | ||
GenBank mRNA | AF021833.1 | - | 9 | 1356 | ||
AK049399.1 | - | 12 | 1971 | |||
BC038483.1 | - | 9 | 1060 | |||
BC054105.1 | - | 9 | 1305 | 30022728 |