Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019930.2 | NP_064314.2 | 3, 4 | 735 | ||
GenBank mRNA | AF006465.1 | - | 4, 5 | 564 | ||
AK132714.1 | - | 3, 4 | 214 |