Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSFA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130445.2 | NP_001123917.1 | 3, 4 | 665 | ||
NM_001287503.1 | NP_001274432.1 | 3, 4 | 665 | |||
NM_006751.6 | NP_006742.2 | 3, 4 | 665 | |||
NR_109843.1 | 3, 4 | 665 | ||||
XM_005246812.1 | XP_005246869.1 | 3, 4 | 573 | |||
XM_005246813.1 | XP_005246870.1 | 3, 4 | 573 | |||
XM_011511701.2 | XP_011510003.1 | 3, 4 | 573 | |||
XM_017004782.1 | XP_016860271.1 | 3, 4 | 573 | |||
GenBank mRNA | AB067514.2 | - | 3, 4 | 513 | ||
AB116937.1 | - | 3, 4 | 380 | |||
AK291745.1 | - | 3, 4 | 401 | |||
BC028706.2 | - | 3, 4 | 426 | 4823606 | ||
BC052581.1 | - | 3, 4 | 434 | 6084501 | ||
BC064499.1 | - | 3, 4 | 434 | 6251533 | ||
BX647257.1 | - | 3, 4 | 396 | |||
BX647967.1 | - | 3, 4 | 298 | |||
BX648888.1 | - | 3, 4 | 186 | |||
CR749488.1 | - | 2, 3 | 410 |