Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177323.2 | NP_001170794.1 | 10 | 1846 | ||
NM_019942.5 | NP_064326.2 | 10 | 1849 | |||
GenBank mRNA | AK048455.1 | - | 10 | 1671 | ||
AK088614.1 | - | 10 | 1612 | |||
AK160884.1 | - | 10 | 1643 |