Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnase2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019957.4 | NP_064341.3 | 4 | 642 | ||
GenBank mRNA | AF128888.1 | - | 3 | 521 | ||
AK079224.1 | - | 4 | 643 | |||
BC075661.1 | - | 3 | 397 | 30606058 |