Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qtnf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204129.1 | NP_001191058.1 | 3 | 293 | ||
NM_001204130.1 | NP_001191059.1 | 3 | 205 | |||
NM_019959.3 | NP_064343.1 | 3 | 283 | |||
GenBank mRNA | AF192499.1 | - | 3 | 231 | ||
AK005484.1 | - | 3 | 252 | |||
AK145104.1 | - | 3 | 206 | |||
BC037636.1 | - | 2 | 198 | |||
BC049894.1 | - | 3 | 234 | 4018549 | ||
CJ056741.1 | - | 3 | 283 | |||
DQ002394.1 | - | 2 | 177 |