Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019960.2 | NP_064344.1 | 1 | 504 | ||
GenBank mRNA | AF203375.1 | - | 1 | 487 | ||
AK009633.1 | - | 1 | 505 | |||
BC065388.1 | - | 1 | 487 | 851802 | ||
BC092376.1 | - | 1 | 474 | 30609467 |