Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164195.1 | NP_001157667.1 | 10 | 1161 | ||
NM_019961.3 | NP_064345.1 | 10 | 1161 | |||
GenBank mRNA | AF152996.1 | - | 10 | 1084 | ||
AK005315.1 | - | 10 | 1017 | |||
AK005978.1 | - | 9 | 858 | |||
AK012827.1 | - | 10 | 1139 | |||
AK075820.1 | - | 10 | 1076 | |||
AK083778.1 | - | 5 | 649 | |||
AK146934.1 | - | 10 | 1140 | |||
BC033415.1 | - | 10 | 1043 | 5008068 | ||
BC037606.1 | - | 10 | 1124 | 5361347 |