Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Miox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019977.2 | NP_064361.2 | 3 | 173 | ||
GenBank mRNA | AF197127.1 | - | 3 | 171 | ||
AK002406.1 | - | 3 | 187 | |||
AK165481.1 | - | 1 | 864 | |||
AY738257.1 | - | 2 | 143 | |||
BC013543.1 | - | 3 | 173 | 4242159 |