Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019981.2 | NP_064365.1 | 6 | 769 | ||
GenBank mRNA | AB022914.1 | - | 6 | 767 | ||
AK005769.1 | - | 6 | 763 | |||
BC048475.1 | - | 6 | 760 | 6740387 |