Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019992.3 | NP_064376.1 | 5 | 510 | ||
GenBank mRNA | AB036058.1 | - | 5 | 500 | ||
AK041474.1 | - | 5 | 511 | |||
AK153661.1 | - | 5 | 589 | |||
AK162655.1 | - | 5 | 592 | |||
BC145550.1 | - | 4 | 412 | 9054114 | ||
BC145828.1 | - | 5 | 525 | 40131131 | ||
BC145830.1 | - | 5 | 526 | 40131133 |