Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh9a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019993.3 | NP_064377.2 | 11 | 2464 | ||
GenBank mRNA | AB296386.1 | - | 11 | 2409 | ||
AK010526.1 | - | 3 | 1017 | |||
AK132336.1 | - | 11 | 2249 | |||
AK154914.1 | - | 11 | 2448 | |||
AK165730.1 | - | 11 | 2466 | |||
AK168878.1 | - | 11 | 2413 | |||
AK208220.1 | - | 1 | 213 | |||
BC003297.1 | - | 11 | 2241 | 3583976 |