Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glmp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020003.1 | NP_064387.1 | 4 | 751 | ||
GenBank mRNA | AB027141.1 | - | 4 | 707 | ||
AB027142.1 | - | 4 | 751 | |||
AK002728.1 | - | 4 | 715 | |||
AK007803.1 | - | 4 | 724 | |||
AK075614.1 | - | 4 | 729 | |||
AK080538.1 | - | 4 | 725 | |||
AK150794.1 | - | 4 | 724 | |||
AK154340.1 | - | 4 | 724 | |||
AK160793.1 | - | 691 | ||||
AK170696.1 | - | 4 | 714 | |||
AK171042.1 | - | 4 | 723 | |||
AK171142.1 | - | 1478 | ||||
AK172606.1 | - | 2 | 1373 | |||
BC021547.1 | - | 4 | 717 | 4507218 |