Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020009.2 | NP_064393.2 | 4 | 433 | ||
GenBank mRNA | AF152838.1 | - | 4 | 408 | ||
AK036004.1 | - | 2 | 1619 | |||
AK172584.1 | - | 4 | 424 | |||
BC021454.1 | - | 4 | 400 | |||
BC043920.1 | - | 4 | 390 | |||
BC112904.1 | - | 4 | 391 | 6465126 |