Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164621.1 | NP_001158093.1 | 6 | 1298 | ||
NM_001164622.1 | NP_001158094.1 | 4 | 833 | |||
NM_020012.2 | NP_064396.2 | 5 | 985 | |||
GenBank mRNA | AF249667.1 | - | 5 | 931 | ||
AF249668.1 | - | 3 | 516 | |||
AK011316.2 | - | 6 | 986 | |||
AK038986.1 | - | 4 | 775 | |||
BC049079.1 | - | 6 | 1298 | |||
BC052205.1 | - | 2994 | ||||
BC054841.1 | - | 5 | 915 | 6489903 | ||
BC094250.1 | - | 5 | 936 | 5720753 |