Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scamp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301634.1 | NP_001288563.1 | 7 | 1450 | ||
NM_001301635.1 | NP_001288564.1 | 6 | 1176 | |||
NM_020270.3 | NP_064666.1 | 7 | 1333 | |||
GenBank mRNA | BC018613.1 | - | 7 | 1297 | 4511417 |