Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3cg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146200.1 | NP_001139672.1 | 2 | 505 | ||
NM_001146201.1 | NP_001139673.1 | 2 | 561 | |||
NM_020272.2 | NP_064668.2 | 2 | 682 | |||
GenBank mRNA | AF208345.1 | - | 1 | 361 | ||
AJ249280.1 | - | 2 | 682 | |||
AK040543.1 | - | 2 | 467 | |||
AK132553.1 | - | 2 | 507 | |||
AK137470.1 | - | 2 | 414 | |||
AK137620.1 | - | 2 | 563 | |||
BC051246.1 | - | 2 | 422 | 6514979 |