Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsmf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039386.1 | NP_001034475.1 | 3 | 590 | ||
NM_001039387.1 | NP_001034476.1 | 3 | 590 | |||
NM_001177654.1 | NP_001171125.1 | 3 | 590 | |||
NM_001177655.1 | NP_001171126.1 | 3 | 590 | |||
NM_020276.3 | NP_064672.2 | 3 | 590 | |||
GenBank mRNA | AF266508.2 | - | 3 | 556 | ||
AJ278902.1 | - | 3 | 368 | |||
AK004986.2 | - | 3 | 587 | |||
AK019560.1 | - | 3 | 557 | |||
AK041478.1 | - | 3 | 567 | |||
AK053402.1 | - | 3 | 537 | |||
AK158803.1 | - | 4 | 389 | |||
AK165300.1 | - | 3 | 535 | |||
BC006642.1 | - | 3 | 548 | 3483536 |