Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsmf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039386.1 | NP_001034475.1 | 4, 5 | 925 | ||
NM_020276.3 | NP_064672.2 | 4, 5 | 925 | |||
GenBank mRNA | AF266508.2 | - | 4 | 891 | ||
AJ278902.1 | - | 4, 5 | 703 | |||
AK019560.1 | - | 4, 5 | 892 | |||
AK158803.1 | - | 5, 6 | 724 |