Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020295.3 | NP_064691.2 | 14 | 1343 | ||
GenBank mRNA | AF190665.1 | - | 14 | 1145 | ||
AK030631.1 | - | 15 | 2149 | |||
BC016110.1 | - | 14 | 1102 | 4481088 |