Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141931.1 | NP_001135403.1 | 9 | 1085 | ||
NM_001141932.1 | NP_001135404.1 | 9 | 1085 | |||
NM_020296.2 | NP_064692.2 | 9 | 1278 | |||
GenBank mRNA | AB026569.1 | - | 9 | 1001 | ||
AK133054.1 | - | 9 | 806 | |||
AK135280.1 | - | 9 | 965 | |||
AK145897.1 | - | 9 | 1087 | |||
AK147109.1 | - | 9 | 1279 | |||
AK161102.1 | - | 9 | 1035 | |||
AK161416.1 | - | 5 | 415 | |||
AK164307.1 | - | 9 | 1079 | |||
AK178547.1 | - | 3 | 116 | |||
AK180025.1 | - | 2 | 117 | |||
AK188295.1 | - | 3 | 116 | |||
AK203574.1 | - | 1 | 32 | |||
AK218973.1 | - | 1 | 24 | |||
BC016501.1 | - | 9 | 1266 | 4527742 | ||
BC056609.1 | - | 6 | 477 | |||
BC057866.1 | - | 9 | 1117 | 5327143 |