Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141931.1 | NP_001135403.1 | 14 | 1640 | ||
NM_001141932.1 | NP_001135404.1 | 15 | 1688 | |||
NM_020296.2 | NP_064692.2 | 14 | 1833 | |||
GenBank mRNA | AA175245.1 | - | 1 | 31 | ||
AB026569.1 | - | 14 | 1556 | |||
AK133054.1 | - | 14 | 1361 | |||
AK135280.1 | - | 14 | 1520 | |||
AK145897.1 | - | 14 | 1642 | |||
AK147109.1 | - | 14 | 1834 | |||
AK159851.1 | - | 2 | 241 | |||
AK161102.1 | - | 15 | 1638 | |||
AK161416.1 | - | 10 | 970 | |||
AK201259.1 | - | 1 | 61 | |||
BC016501.1 | - | 14 | 1821 | 4527742 | ||
BC056609.1 | - | 12 | 1080 | |||
BC057866.1 | - | 15 | 1720 | 5327143 |