Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020330.4 | NP_065063.1 | 2 | 978 | ||
GenBank mRNA | AF251559.1 | - | 1 | 470 | ||
AK014827.2 | - | 2 | 979 | |||
BC132152.1 | - | 1 | 521 | 40130429 | ||
BC132344.1 | - | 1 | 521 | 40130621 |