Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289757.1 | NP_001276686.1 | 10 | 1141 | ||
NM_021470.6 | NP_067445.3 | 10 | 1113 | |||
GenBank mRNA | AF190664.1 | - | 7 | 875 | ||
AK005795.1 | - | 10 | 1068 | |||
AK006379.1 | - | 10 | 1057 | |||
AK012288.1 | - | 8 | 873 | |||
BC050964.1 | - | 10 | 1056 | |||
BC061103.1 | - | 9 | 714 | |||
BC089493.1 | - | 10 | 1056 | 6741898 |