Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021546.3 | NP_067521.2 | 13 | 1504 | ||
GenBank mRNA | AB039948.1 | - | 11 | 1325 | ||
AK013520.1 | - | 12 | 1431 | |||
AK044296.1 | - | 13 | 1496 | |||
AK158225.1 | - | 13 | 1506 |