Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgcr14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081633.1 | NP_001075102.1 | 2 | 282 | ||
NM_022408.2 | NP_071853.2 | 2 | 282 | |||
GenBank mRNA | AF037256.1 | - | 2 | 281 | ||
AK088520.1 | - | 2 | 247 | |||
AK142684.1 | - | 2 | 247 | |||
AK148404.1 | - | 2 | 250 | |||
AK177724.1 | - | 2 | 125 | |||
AK209733.1 | - | 1 | 103 | |||
BC013711.1 | - | 2 | 261 | 3708646 | ||
BY069641.1 | - | 2 | 283 |