Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022653.4 | NP_073144.3 | 12 | 2032 | ||
GenBank mRNA | AF314187.1 | - | 12 | 1990 | ||
AK028609.1 | - | 12 | 1966 | |||
BC031175.1 | - | 4 | 543 | |||
BC031722.1 | - | 12 | 1998 | 4019169 | ||
BC039651.1 | - | 4 | 543 |