Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023141.2 | NP_075630.2 | 6 | 2276 | ||
GenBank mRNA | AJ299404.1 | - | 6 | 2242 | ||
BC052851.1 | - | 6 | 2561 |