Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tollip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023764.3 | NP_076253.1 | 6 | 1313 | ||
GenBank mRNA | AK015062.1 | - | 1256 | |||
AK016482.1 | - | 6 | 1314 | |||
AK045422.1 | - | 6 | 1297 | |||
AK049263.1 | - | 6 | 1335 | |||
AK049394.1 | - | 6 | 1259 | |||
AK161312.1 | - | 6 | 1300 | |||
BC053515.1 | - | 6 | 1300 | |||
BC062139.1 | - | 6 | 1294 | 6334946 |